missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl p-toluenesulfonate, 97%

Product Code. 11423834
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423834 500 g 500g
11403834 25 g 25g
11413834 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423834 Supplier Thermo Scientific Alfa Aesar Supplier No. A11863.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 80-41-1 | C9H11ClO3S | 234.694 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-41-1
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.694
MDL Number MFCD00000970
InChI Key ZXNMIUJDTOMBPV-UHFFFAOYSA-N
Synonym 2-chloroethyl p-toluenesulfonate, 2-chloroethyl tosylate, 2-chloroethyl p-tosylate, 2-tosyloxyethyl chloride, 2-chloroethanol p-toluenesulfonate, p-toluenesulfonic acid, 2-chloroethyl ester, ethanol, 2-chloro-, 4-methylbenzenesulfonate, 2-p-toluenesulfonyloxy ethyl chloride, 2-chloroethyl-p-toluenesulfonate, p-toluenesulfonic acid-2-chloroethyl ester
PubChem CID 6639
IUPAC Name 2-chloroethyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCCl

Specifications

Density 1.294
Boiling Point 158°C to 159°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.529
Quantity 500 g
Beilstein 1968816
Formula Weight 234.7
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl p-toluenesulfonate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.