missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methyl ether, 98%

Product Code. 11471077
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471077 500 g 500g
11451077 25 g 25g
11461077 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471077 Supplier Thermo Scientific Alfa Aesar Supplier No. B24081.36
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 627-42-9 | C3H7ClO | 94.54 g/mol

2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethyl methyl ether was used in the synthesis of acyclic nucleosides of thieno[2,3-d] pyrimidine derivatives.

Solubility
Soluble in water 60 g/L at 20°C.

Notes
Store away from oxidizing agents, bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-42-9
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00000972
InChI Key XTIGGAHUZJWQMD-UHFFFAOYSA-N
Synonym 2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
PubChem CID 12316
IUPAC Name 1-chloro-2-methoxyethane
SMILES COCCCl

Specifications

Density 1.04
Boiling Point 89°C to 90°C
Flash Point 8°C (46°F)
Linear Formula CH3OCH2CH2Cl
Refractive Index 1.409
Quantity 500 g
UN Number UN3271
Solubility Information Soluble in water 60g/L at 20°C.
Formula Weight 94.54
Percent Purity 98%
Chemical Name or Material 2-Chloroethyl methyl ether
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.