missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl methyl ether, 98%

Product Code. 10286750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Unit Size:
100mL
2.5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10286750 500 mL 500mL
10042900 100 mL 100mL
10379033 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10286750 Supplier Thermo Scientific Acros Supplier No. 221775000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 627-42-9 | C3H7ClO | 94.54 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 627-42-9
Molecular Formula C3H7ClO
Molecular Weight (g/mol) 94.54
MDL Number MFCD00000972
InChI Key XTIGGAHUZJWQMD-UHFFFAOYSA-N
Synonym 2-chloroethyl methyl ether, 2-methoxyethyl chloride, ethane, 1-chloro-2-methoxy, 2-chloroethylmethylether, unii-u890tj10ym, chloro-2-methoxyethane, chloroethyl-methyl ether, 1-chloro-2-methoxy-ethane, ether, 2-chloroethyl methyl
PubChem CID 12316
IUPAC Name 1-chloro-2-methoxyethane
SMILES COCCCl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -55°C
Color Colorless to Yellow
Density 1.0300g/mL
Boiling Point 89°C to 90°C
Flash Point 15°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CH2OCH3
Refractive Index 1.4080 to 1.41
Quantity 500 mL
Beilstein 01,337
Specific Gravity 1.03
Solubility Information Solubility in water: 60g/L (20 c)
Viscosity 0.65 mPa.s (20°C)
Formula Weight 94.54
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl methyl ether, 98%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethyl methyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P403+P235-Store in a well-ventilated place. Keep cool.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.