missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl isothiocyanate, 97%

Product Code. 11372517
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11372517 1 g 1g
11382517 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11372517 Supplier Thermo Scientific Alfa Aesar Supplier No. L11604.03
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 6099-88-3 | C3H4ClNS | 121.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6099-88-3
Molecular Formula C3H4ClNS
Molecular Weight (g/mol) 121.582
MDL Number MFCD00041125
InChI Key ZUWFBQUHBOUPFK-UHFFFAOYSA-N
Synonym 2-chloroethyl isothiocyanate, 2-chloroethylisothiocyanate, ethane, 1-chloro-2-isothiocyanato, sgxdlhbabrkth@, chloroethylisothiocyanate, acmc-1b2t4, beta-chloroethylisothiocyanate, beta-chloroethyl isothiocyanate, 1-chloro-2-isothiocyanato-ethane, #
PubChem CID 138661
IUPAC Name 1-chloro-2-isothiocyanatoethane
SMILES C(CCl)N=C=S

Specifications

Density 1.265
Boiling Point 79°C to 80°C (13 mmHg)
Flash Point 96°C (205°F)
Refractive Index 1.556
Quantity 1 g
UN Number UN2922
Beilstein 605585
Sensitivity Moisture sensitive
Formula Weight 121.59
Percent Purity 97%
Chemical Name or Material 2-Chloroethyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.