missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl chloroformate, 97%

Product Code. 11944941 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11944941 100 g Glass bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11944941 Supplier Thermo Scientific Acros Supplier No. 395331000
 Restricted Item
Estimated Shipment: 18-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 627-11-2 | C3H4Cl2O2 | 142.96 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Specifications

CAS 627-11-2
CAS Min % 96.0
CAS Max % 100.0
Melting Point -70.0°C
Color Colorless to Yellow
Density 1.3900g/mL
Boiling Point 154.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C3H4Cl2O2
Linear Formula ClCH2CH2OCOCl
Refractive Index 1.4450 to 1.4470
MDL Number MFCD00000646
Quantity 100 g
Specific Gravity 1.39
Synonym 2-chloroethyl chloroformate, chloroethyl chloroformate, carbonochloridic acid, 2-chloroethyl ester, chloroformic acid 2-chloroethyl ester, formic acid, chloro-, 2-chloroethyl ester, chloroethylchloroformate, 2-chloroethylchloroformate, unii-4d32h07z9i, beta-chloroethyl chlorocarbonate, chloroformic acid, beta-chloroethyl ester
Solubility Information Solubility in water: hydrolyzes
InChI Key SVDDJQGVOFZBNX-UHFFFAOYSA-N
SMILES ClCCOC(Cl)=O
IUPAC Name 2-chloroethyl carbonochloridate
Molecular Weight (g/mol) 142.96
PubChem CID 12305
Formula Weight 142.97
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chloroethyl chloroformate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethyl chloroformate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H330-Fatal if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.