missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl 4-methylbenzenesulfonate, TRC

Product Code. 30357789
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357789 25 g -
30355450 1 g 1g
30350936 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357789 Supplier Toronto Research Chemicals Supplier No. TRCC36321525G
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 80-41-1
Molecular Formula C9H11ClO3S
Molecular Weight (g/mol) 234.7
Synonym 2-(p-Toluenesulfonyloxy)ethyl chloride, 2-Chloro-1-(p-toluenesulfonyl)ethane, 2-Chloroethanol p-toluenesulfonate, 2-Chloroethanol tosylate, 2-Chloroethyl p-toluenesulfonate, 2-Chloroethyl p-tosylate, 2-Chloroethyl tosylate, 2-Tosyloxyethyl chloride, p-Toluenesulfonic acid 2-chloroethyl ester, beta-Chloroethyl p-toluenesulfonate, beta-Chloroethyl tosylate
IUPAC Name 2-chloroethyl 4-methylbenzenesulfonate
SMILES Cc1ccc(cc1)S(=O)(=O)OCCCl

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H11ClO3S/c1-8-2-4-9(5-3-8)14(11,12)13-7-6-10/h2-5H,6-7H2,1H3
Formula Weight 234.0117
Chemical Name or Material 2-Chloroethyl 4-methylbenzenesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.