missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethyl (2-Chloroethyl)phosphonate, TRC

Product Code. 30133901
Change view
Click to view available options
Quantity:
100 mg
1 g
Unit Size:
100mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133901 1 g 1g
30006607 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30133901 Supplier Toronto Research Chemicals Supplier No. TRCC3660651G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17378-30-2
Molecular Formula C4 H9 Cl2 O3 P
Molecular Weight (g/mol) 206.99
Synonym Mono-2-chloroethyl 2-Chloroethylphosphonate, O-(2-Chloroethyl) Hydrogen 2-Chlorethylphosphonate, 2-Chloroethanephosphonic Acid Mono-2-chloroethyl Ester, 2-Chloro (2-Chloroethyl) Hydrogen Phosphonate Ethanol;
IUPAC Name 2-chloroethoxy(2-chloroethyl)phosphinic acid
SMILES OP(=O)(CCCl)OCCCl

Specifications

Quantity 1 g
InChI Formula InChI=1S/C4H9Cl2O3P/c5-1-3-9-10(7,8)4-2-6/h1-4H2,(H,7,8)
Formula Weight 205.9666
Percent Purity >95%
Chemical Name or Material 2-Chloroethyl (2-Chloroethyl)phosphonate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.