missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanol, 99%, Thermo Scientific Chemicals

Product Code. 15481457
Change view
Click to view available options
Quantity:
50 mL
250 mL
1 L
Unit Size:
1 L
250mL
50mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15481457 50 mL 50mL
15491457 250 mL 250mL
15401467 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15481457 Supplier Thermo Scientific Alfa Aesar Supplier No. 036636.AD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

2-Chloroethanol is used in the preparation of several dyes by the alkylation of aniline derivatives with chloroethanol. It is a building block in the production of pharmaceuticals and plasticizers. Also used for manufacture of thiodiglycol. It is a solvent for cellulose acetate and ethyl cellulose, textile printing dyes, in dewaxing, refining of rosin, extraction of pine lignin and the cleaning of machines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-07-3
Molecular Formula C2H5ClO
Molecular Weight (g/mol) 80.511
MDL Number MFCD00002829
InChI Key SZIFAVKTNFCBPC-UHFFFAOYSA-N
Synonym ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
PubChem CID 34
ChEBI CHEBI:28200
IUPAC Name 2-chloroethanol
SMILES C(CCl)O

Specifications

Melting Point -63°C
Color Yellow
Density 1.202 g/mL
Boiling Point 129°C
Flash Point 55°C (131°F)
Odor Ether-like
Linear Formula ClCH2CH2OH
Refractive Index 1.441
Quantity 50 mL
UN Number UN1135
Beilstein 878139
Merck Index 14,3793
Solubility Information Fully miscible with water.
Formula Weight 80.51
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.