missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanol, 99+%, Thermo Scientific Chemicals

Catalog Number 10367600
Click to view available options
Quantity:
100 mL
1 L
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
107-07-3
80.51
MFCD00002829
SZIFAVKTNFCBPC-UHFFFAOYSA-N
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
34
CHEBI:28200
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
107-07-3
80.51
MFCD00002829
SZIFAVKTNFCBPC-UHFFFAOYSA-N
ethylene chlorohydrin, ethanol, 2-chloro, glycol chlorohydrin, 2-chloroethyl alcohol, chloroethanol, glycol monochlorohydrin, 2-monochloroethanol, ethylchlorohydrin, 2-chlorethanol, 2-chloro-1-ethanol
34
CHEBI:28200

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Acidity 0.2mg KOH/g max.
CAS Min % 99.0
CAS Max % 100.0
Melting Point -63.0°C
Color Colorless
Density 1.2000g/mL
Boiling Point 129.0°C
Flash Point 55°C
Infrared Spectrum Authentic
Assay Percent Range 99% min. (GC)
Packaging Glass bottle
Molecular Formula C2H5ClO
Linear Formula ClCH2CH2OH
Refractive Index 1.4405 to 1.4425
Quantity 100 mL
Beilstein 01, 337
Fieser 05,117
Merck Index 15, 3847
Specific Gravity 1.2
Solubility Information Solubility in water: miscible. Other solubilities: miscible with alcohol,soluble in most organic liquids,insoluble in alifatic hydrocarbons
SMILES C(CCl)O
IUPAC Name 2-chloroethanol
Viscosity 3.4 mPa.s (20°C)
Formula Weight 80.51
Percent Purity 99+%
Physical Form Liquid
Chemical Name or Material 2-Chloroethanol, 99+%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroethanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 2
  • Acute toxicity Category 1
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H330-Fatal if inhaled.
  • H310-Fatal in contact with skin.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P361-Take off immediately all contaminated clothing.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'