missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroethanesulfonyl chloride, 98%

Product Code. 11359639
Click to view available options
Quantity:
10 g
50 g
100 g
250 g
Unit Size:
100g
10g
250g
50g
This item is not returnable. View return policy

Product Code. 11359639

Brand: Thermo Scientific Alfa Aesar A15377.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1622-32-8 | C2H4Cl2O2S | 163.012 g/mol

2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1622-32-8
Molecular Formula C2H4Cl2O2S
Molecular Weight (g/mol) 163.012
MDL Number MFCD00007461
InChI Key VHCSBTPOPKFYIU-UHFFFAOYSA-N
Synonym 2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
PubChem CID 15385
IUPAC Name 2-chloroethanesulfonyl chloride
SMILES C(CCl)S(=O)(=O)Cl

Specifications

Density 1.56
Boiling Point 203°C
Flash Point >110°C (230°F)
Linear Formula ClCH2CH2SO2Cl
Refractive Index 1.4915
Quantity 100 g
UN Number UN3390
Beilstein 1751202
Sensitivity Moisture sensitive
Solubility Information Reacts with water.
Formula Weight 163.02
Percent Purity 98%
Chemical Name or Material 2-Chloroethanesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.