missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorocyclohexanone, 95%, stabilized

Product Code. 10438250 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10438250 100 g Plastic bottle 100g
10752505 5 g Plastic bottle 5g
10478440 25 g Plastic bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10438250 Supplier Thermo Scientific Acros Supplier No. 108961000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 822-87-7 | C6H9ClO | 132.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 822-87-7
Molecular Formula C6H9ClO
Molecular Weight (g/mol) 132.59
MDL Number MFCD00001626
InChI Key CCHNWURRBFGQCD-UHFFFAOYNA-N
Synonym 2-chlorocyclohexanone, cyclohexanone, 2-chloro, alpha-chlorocyclohexanone, 2-chlorocyclohexonone, .alpha.-chlorocyclohexanone, 2-chlorocyclohexanon, 2-chlorocylohexanone, 2-chloro cyclohexanone, 2-chloro-cyclohexanone, acmc-209ted
PubChem CID 13203
IUPAC Name 2-chlorocyclohexan-1-one
SMILES ClC1CCCCC1=O

Specifications

CAS Min % 94.0
CAS Max % 100.0
Melting Point 23°C
Color Brown
Density 1.1600g/mL
Boiling Point 82°C to 83°C (10.0 mmHg)
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Plastic bottle
Linear Formula ClC6H9(=O)
Refractive Index 1.4830 to 1.485
Quantity 100 g
Beilstein 07, 10
Specific Gravity 1.16
Stabilizer 0.2% CaCO3/MgO (1:1)
Formula Weight 132.59
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 2-Chlorocyclohexanone, 95%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorocyclohexanone, stabilized
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.