missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 2-Chlorobutyryl chloride, tech. 85%

Catalog Number p-7058366
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
7623-11-2
MFCD00041409
This item is not returnable. View return policy
7623-11-2
MFCD00041409
Applications
2-Chlorobutyryl chloride is used as a pharmaceutical intermediate.

Solubility
Reacts with water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents. It is sensitive to moisture.
TRUSTED_SUSTAINABILITY
Density 1.236
Boiling Point 138°C to 140°C
Molecular Formula C12H13NO2S
Refractive Index 1.451
Quantity 5 g
UN Number UN3265
Beilstein 741947
Sensitivity Moisture sensitive
Synonym ethyl 3-methyl-4h-benzo b 1,4 thiazine-2-carboxylate, ethyl 3-methyl-4h-benzo e 1,4-thiazine-2-carboxylate, 3-methyl-4h-benzo 1,4 thiazine-2-carboxylic acid ethyl ester, maybridge1_006434, cbmicro_028856, 2-carbethoxy-3-methyl-4h-1,4-benzothiazine, 3-methyl-4h-1,4-benzothiazine-2-carboxylic acid ethyl ester, 4h-1,4-benzothiazine-2-carboxylicacid, 3-methyl-, ethyl ester, 4h-1,4-benzothiazine-2-carboxylicacid,3-methyl-,ethyl ester
Solubility Information Reacts with water.
InChI Key NCZIRXVLKKERGP-UHFFFAOYSA-N
SMILES CCOC(=O)C1=C(NC2=CC=CC=C2S1)C
IUPAC Name ethyl 3-methyl-4H-1,4-benzothiazine-2-carboxylate
Molecular Weight (g/mol) 235.30
PubChem CID 360308
Formula Weight 141
Percent Purity 85%
Grade Technical
Chemical Name or Material 2-Chlorobutyryl chloride
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'