missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobutane, 98+%

Product Code. p-7025871
Click to view available options
Quantity:
50 mL
250 mL
1000 mL
Unit Size:
1000mL
250mL
50mL
This item is not returnable. View return policy

Product Code. 11369639

Brand: Thermo Scientific Alfa Aesar B21586.AD

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 78-86-4 | C4H9Cl | 92.566 g/mol

2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chlorobutane is used as a solvent and intermediate in organic synthesis. Further, it is involved in the preparation of active pharmaceutical ingredients, plasticizer, rubber, resin and surfactants.

Solubility
Immiscible with water.

Notes
Incompatible with metals, copper, strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 78-86-4
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.566
MDL Number MFCD00000871
InChI Key BSPCSKHALVHRSR-UHFFFAOYSA-N
Synonym sec-butyl chloride, butane, 2-chloro, 1-methylpropyl chloride, 2-chloro-butane, ccris 9391, alkanes, c11-14, 2-chloro, sec.-butyl chloride, sjphadilup@, sec-c4h9cl, acmc-209pfl
PubChem CID 6563
IUPAC Name 2-chlorobutane
SMILES CCC(C)Cl

Specifications

Melting Point -141°C to -140°C
Density 0.873
Boiling Point 67°C to 69°C
Flash Point −15°C (5°F)
Assay Percent Range 98+%
Odor Sweet
Refractive Index 1.397
Quantity 50 mL
UN Number UN1127
Beilstein 1718770
Merck Index 14,1561
Solubility Information Immiscible with water.
Formula Weight 92.57
Percent Purity ≥98%
Physical Form Liquid
Chemical Name or Material 2-Chlorobutane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.