missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzylamine, 96%

Product Code. 11467447
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11467447 500 g 500g
11447447 25 g 25g
11457447 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11467447 Supplier Thermo Scientific Alfa Aesar Supplier No. A15594.36
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 89-97-4 | C7H8ClN | 141.598 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89-97-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.598
MDL Number MFCD00008108
InChI Key KDDNKZCVYQDGKE-UHFFFAOYSA-N
Synonym 2-chlorobenzylamine, 2-chlorophenyl methanamine, o-chlorobenzylamine, benzenemethanamine, 2-chloro, o-chlorobenzyl amine, 2-chlorobenzenemethanamine, 2-chloro-benzylamine, benzylamine, o-chloro, 2-chlorophenyl methylamine, unii-l790j4y7a3
PubChem CID 66648
IUPAC Name (2-chlorophenyl)methanamine
SMILES C1=CC=C(C(=C1)CN)Cl

Specifications

Density 1.17
Boiling Point 227°C
Flash Point 88°C (190°F)
Odor Amine-like
Refractive Index 1.562
Quantity 500 g
UN Number UN2735
Beilstein 907186
Sensitivity Air Sensitive
Formula Weight 141.6
Percent Purity 96%
Chemical Name or Material 2-Chlorobenzylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.