missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzyl isothiocyanate, 97%
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Description
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
2-Chlorobenzyl isothiocyanate is used as pharmaceutical intermediate.
Solubility
It hydrolyzes in water.
Notes
Moisture sensitive. Incompatible with oxidizing agents, alcohols and amines.
Specifications
Specifications
| Density | 1.149 |
| Boiling Point | 137°C to 138°C (6 mmHg) |
| Quantity | 1 g |
| Odor | Irritating |
| Refractive Index | 1.614 |
| UN Number | UN2922 |
| Beilstein | 2936688 |
| Solubility Information | It hydrolyzes in water. |
| Formula Weight | 183.66 |
| Percent Purity | 97% |
| Show More |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction