missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl Cyanide, TRC

Product Code. 30352295
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352295 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352295 Supplier Toronto Research Chemicals Supplier No. TRCC3644651G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chlorobenzyl Cyanide
CAS 2856-63-5
Molecular Formula C8 H6 Cl N
Quantity 1 g
Synonym 2-Chloro-benzeneacetonitrile, (o-Chlorophenyl)-acetonitrile, (2-Chlorophenyl)acetonitrile, 2-Chlorobenzeneacetonitrile, NSC 39462, o-Chlorobenzyl Cyanide, o-Chlorobenzyl Nitrile, o-Chlorophenylacetonitrile
InChI Formula InChI=1S/C8H6ClN/c9-8-4-2-1-3-7(8)5-6-10/h1-4H,5H2
SMILES Clc1ccccc1CC#N
IUPAC Name 2-(2-chlorophenyl)acetonitrile
Molecular Weight (g/mol) 151.59
Formula Weight 151.0189
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.