missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl chloride, 98+%

Product Code. 10134300 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Packaging:
Glass bottle
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10134300 250 mL Glass bottle 250mL
10295900 1 L Glass bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10134300 Supplier Thermo Scientific Acros Supplier No. 186532500
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 611-19-8 | C7H6Cl2 | 161.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-19-8
Molecular Formula C7H6Cl2
Molecular Weight (g/mol) 161.03
MDL Number MFCD00000893
InChI Key BASMANVIUSSIIM-UHFFFAOYSA-N
Synonym 2-chlorobenzyl chloride, 1-chloro-2-chloromethyl benzene, 2-chlorobenzylchloride, o-chlorobenzyl chloride, alpha,2-dichlorotoluene, benzene, 1-chloro-2-chloromethyl, alpha,o-dichlorotoluene, ar,alpha-dichlorotoluene, ortho-alpha-dichlorotoluene, unii-h58di83rmd
PubChem CID 11906
IUPAC Name 1-chloro-2-(chloromethyl)benzene
SMILES C1=CC=C(C(=C1)CCl)Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Melting Point -13°C
Color Colorless to Yellow
Density 1.2700g/mL
Boiling Point 213°C to 214°C
Flash Point 82°C
Infrared Spectrum Authentic
Assay Percent Range 98% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2Cl
Refractive Index 1.5580 to 1.56
Quantity 250 mL
Beilstein 05, 297
Specific Gravity 1.27
Solubility Information Solubility in water: insoluble. Other solubilities: insoluble in most common solvents (reacts with man
Formula Weight 161.03
Percent Purity 98+%
Physical Form Liquid
Chemical Name or Material 2-Chlorobenzyl chloride, 98+%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chlorobenzyl chloride
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.