missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl bromide, 97%

Product Code. 11410083
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11410083 100 g 100g
11490073 5 g 5g
11400083 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11410083 Supplier Thermo Scientific Alfa Aesar Supplier No. A12129.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 611-17-6 | C7H6BrCl | 205.479 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 611-17-6
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00000566
InChI Key PURSZYWBIQIANP-UHFFFAOYSA-N
Synonym 2-chlorobenzyl bromide, 1-bromomethyl-2-chlorobenzene, o-chlorobenzyl bromide, 2-chlorobenzylbromide, benzene, 1-bromomethyl-2-chloro, alpha-bromo-2-chlorotoluene, unii-cev23h8uv4, 2chlorobenzyl bromide, alpha-bromo-o-chlorotoluene, cev23h8uv4
PubChem CID 11905
IUPAC Name 1-(bromomethyl)-2-chlorobenzene
SMILES C1=CC=C(C(=C1)CBr)Cl

Specifications

Density 1.583
Boiling Point 103°C to 104°C (10 mmHg)
Flash Point 108°C (226°F)
Odor Pungent
Refractive Index 1.592
Quantity 100 g
UN Number UN3265
Beilstein 386264
Formula Weight 205.49
Percent Purity 97%
Chemical Name or Material 2-Chlorobenzyl bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.