missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl alcohol, 99%

Product Code. 10174270 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10174270 50 g 50g
10327820 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10174270 Supplier Thermo Scientific Acros Supplier No. 163420500
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17849-38-6 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17849-38-6
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00004604
InChI Key MBYQPPXEXWRMQC-UHFFFAOYSA-N
Synonym 2-chlorobenzyl alcohol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, 2-chlorobenzenemethanol, o-chlorobenzyl alcohol, benzyl alcohol, o-chloro, 2-chloro-phenyl-methanol, 2-chlorophenyl methan-1-ol, benzenemethanol, ar-chloro, 2-chlorobenzylalcohol
PubChem CID 28810
IUPAC Name (2-chlorophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 69°C to 72°C
Color White
Boiling Point 227°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4CH2OH
Quantity 50 g
Beilstein 06, 444
Solubility Information Solubility in water: limited soluble in water. Other solubilities: 1g/10mL in methanol
Formula Weight 142.58
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 2-Chlorobenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.