missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzyl alcohol, 98+%

Product Code. 11420254
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420254 100 g 100g
11410254 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11420254 Supplier Thermo Scientific Alfa Aesar Supplier No. A13657.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 17849-38-6 | C7H7ClO | 142.582 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17849-38-6
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.582
MDL Number MFCD00004604
InChI Key MBYQPPXEXWRMQC-UHFFFAOYSA-N
Synonym 2-chlorobenzyl alcohol, 2-chlorophenyl methanol, benzenemethanol, 2-chloro, 2-chlorobenzenemethanol, o-chlorobenzyl alcohol, benzyl alcohol, o-chloro, 2-chloro-phenyl-methanol, 2-chlorophenyl methan-1-ol, benzenemethanol, ar-chloro, 2-chlorobenzylalcohol
PubChem CID 28810
IUPAC Name (2-chlorophenyl)methanol
SMILES C1=CC=C(C(=C1)CO)Cl

Specifications

Melting Point 69°C to 71°C
Boiling Point 227°C
Quantity 100 g
Beilstein 2042182
Formula Weight 142.59
Percent Purity ≥98%
Chemical Name or Material 2-Chlorobenzyl alcohol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.