missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chlorobenzoyl Chloride, TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | 2-Chlorobenzoyl Chloride |
| CAS | 609-65-4 |
| Molecular Formula | C7H4Cl2O |
| Quantity | 50 g |
| Synonym | NSC 93897, o-Chlorobenzoyl Chloride |
| InChI Formula | InChI=1S/C7H4Cl2O/c8-6-4-2-1-3-5(6)7(9)10/h1-4H |
| SMILES | ClC(=O)c1ccccc1Cl |
| IUPAC Name | 2-chlorobenzoyl chloride |
| Molecular Weight (g/mol) | 175.01 |
| Formula Weight | 173.9639 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction