missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic hydrazide, 97%, Thermo Scientific™

Catalog Number 10405682
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
5814-05-1
C7H7ClN2O
170.60
MFCD00007597
KPPNLSKVTKSSTG-UHFFFAOYSA-N
2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
79883
2-chlorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)Cl
This item is not returnable. View return policy
5814-05-1
C7H7ClN2O
170.60
MFCD00007597
KPPNLSKVTKSSTG-UHFFFAOYSA-N
2-chlorobenzhydrazide, 2-chlorobenzoic hydrazide, 2-chlorobenzoic acid hydrazide, 2-chlorobenzene-1-carbohydrazide, benzoylhydrazine, o-chloro, benzoic acid, 2-chloro-, hydrazide, 2-chlorobenzenecarbohydrazide, chlorobenzohydrazide, o-chlorobenzhydrazide, 2clphcon2
79883
2-chlorobenzohydrazide
C1=CC=C(C(=C1)C(=O)NN)Cl
CAS Min % 96.0
CAS Max % 100.0
Melting Point 113°C to 118°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula ClC6H4CONHNH2
Quantity 25 g
Formula Weight 170.6
Percent Purity ≥96.0%
Physical Form Crystalline Powder
Chemical Name or Material 2-Chlorobenzoic hydrazide, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'