missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzoic acid, 98%

Product Code. 10657314 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
2 kg
Packaging:
Glass Bottle
Plastic bottle
Unit Size:
100g
2kg
500g
5g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10657314 2 kg Plastic bottle 2kg
10123690 5 g Glass Bottle 5g
10091980 100 g Plastic bottle 100g
10002000 500 g Plastic bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 10657314 Supplier Thermo Scientific Acros Supplier No. undefined
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 118-91-2 | C7H5ClO2 | 156.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 118-91-2
Molecular Formula C7H5ClO2
Molecular Weight (g/mol) 156.57
MDL Number MFCD00002412
InChI Key IKCLCGXPQILATA-UHFFFAOYSA-N
Synonym o-chlorobenzoic acid, benzoic acid, o-chloro, benzoic acid, 2-chloro, 2-cba, kyselina o-chlorbenzoova, chlorobenzoic acid, ocba, benzoic acid, chloro, 2-chloro-benzoic acid, chlorobenzoate
PubChem CID 8374
ChEBI CHEBI:30793
IUPAC Name 2-chlorobenzoic acid
SMILES C1=CC=C(C(=C1)C(=O)O)Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 139.0°C to 143.0°C
Color White
Boiling Point 285.0°C
Flash Point 173°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Plastic bottle
Linear Formula ClC6H4CO2H
Quantity 2 kg
Beilstein 09, 334
Merck Index 15, 2125
Solubility Information Solubility in water: soluble in hot water. Other solubilities: freely soluble in alcohol and ether
Formula Weight 156.57
Percent Purity 98%
Physical Form Powder
Chemical Name or Material 2-Chlorobenzoic acid, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chlorobenzoic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.