Learn More
2-Chlorobenzimidazole, 97%
Description
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chlorobenzimidazole is mainly used for the synthesis of medicine and as an intermediate in organic synthesis.
Solubility
Insoluble in water.
Notes
Store in cool, dry place, in well sealed container. Store away from oxidizing agents, alkali metals, bases.
Specifications
Specifications
| Melting Point | ∼180°C to 185°C |
| Molecular Formula | C7H5ClN2 |
| Quantity | 25 g |
| Beilstein | 116526 |
| Synonym | 2-chlorobenzimidazole, 2-chloro-1h-benzo d imidazole, 1h-benzimidazole, 2-chloro, 2-chloro-1h-benzoimidazole, 2-chloro-1h-1,3-benzodiazole, benzimidazole, 2-chloro, 2-chloro-1h-1,3-benzimidazole, 2-chlorobenzoimidazole, pubchem7598, 2-chloro benzimidazole |
| Solubility Information | Insoluble in water. |
| InChI Key | AYPSHJCKSDNETA-UHFFFAOYSA-N |
| SMILES | ClC1=NC2=CC=CC=C2N1 |
| IUPAC Name | 2-chloro-1H-benzimidazole |
| Molecular Weight (g/mol) | 152.58 |
| Show More |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.