missing translation for 'onlineSavingsMsg'
Learn More

2-Chlorobenzenesulfonyl chloride, 98%

Product Code. 10127893 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10127893 25 g Glass bottle 25g
10028691 1 g Glass bottle 1g
10722633 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10127893 Supplier Thermo Scientific Acros Supplier No. 357460250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2905-23-9 | C6H4Cl2O2S | 211.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-23-9
Molecular Formula C6H4Cl2O2S
Molecular Weight (g/mol) 211.06
MDL Number MFCD00051520
InChI Key KMVZDSQHLDGKGV-UHFFFAOYSA-N
Synonym 2-chlorobenzenesulphonyl chloride, 2-chlorobenzene-1-sulfonyl chloride, 2-chlorobenzenesulfonylchloride, benzenesulfonyl chloride, 2-chloro, o-chlorobenzenesulfonyl chloride, o-chlorobenzenesulfochloride, 2-chlorophenylsulfonyl chloride, 2-chloro-benzenesulfonyl chloride, benzenesulfonyl chloride, chloro, chloro 2-chlorophenyl sulfone
PubChem CID 76187
SMILES ClC1=CC=CC=C1S(Cl)(=O)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Orange-Red to Yellow
Density 1.5480g/mL
Boiling Point 150.0°C to 152.0°C (12.0 mmHg)
Flash Point 60°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula ClC6H4SO2Cl
Refractive Index 1.5745 to 1.5765
Quantity 25 g
Specific Gravity 1.548
IUPAC Name 2-chlorobenzenesulfonyl chloride
Formula Weight 211.07
Percent Purity 98%
Physical Form Liquid or Low Melting Solid
Chemical Name or Material 2-Chlorobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.