missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline, TRC

Product Code. 30360824
Change view
Click to view available options
Quantity:
150 mL
500 mL
Unit Size:
150mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30360824 500 mL 500mL
30353227 150 mL 150mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30360824 Supplier Toronto Research Chemicals Supplier No. TRCC364370500ML

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6 H6 Cl N
Molecular Weight (g/mol) 127.57
Synonym Aniline, o-chloro- (8CI), 2-Chlorobenzenamine, 1-Amino-2-chlorobenzene, 2-Amino-1-chlorobenzene, 2-Chloroaniline, 2-Chlorophenylamine, Fast Yellow GC Base, NSC 6183, o-Aminochlorobenzene, o-Chloroaniline
IUPAC Name 2-chloroaniline
SMILES Nc1ccccc1Cl

Specifications

Quantity 500 mL
InChI Formula InChI=1S/C6H6ClN/c7-5-3-1-2-4-6(5)8/h1-4H,8H2
Formula Weight 127.0189
Chemical Name or Material 2-Chloroaniline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.