missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline hydrochloride, 97%

Product Code. p-7046216
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11363097

Brand: Thermo Scientific Alfa Aesar L13483.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 137-04-2 | C6H7Cl2N | 164.03 g/mol

2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline hydrochloride, is used as ice dye basement, used for towels, colored yarn and cloth of cotton, viscose fabric dyeing and printing of chromogenic agent. It is also used as an intermediate in organic synthesis.

Solubility
It is soluble in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 137-04-2
Molecular Formula C6H7Cl2N
Molecular Weight (g/mol) 164.03
MDL Number MFCD00053282
InChI Key DRGIDRZFKRLQTE-UHFFFAOYSA-N
Synonym 2-chloroaniline hydrochloride, 2-chloroanilinium chloride, benzenamine, 2-chloro-, hydrochloride, benzenamine, 2-chloro-, hydrochloride 1:1, 2-chlorophenylamine, chloride, acmc-209cai, o-chloroaniline hydrochloride, 95-51-2 parent, 2-chloranylaniline hydrochloride, benzenamine,2-chloro-,hydrochloride 1:1
PubChem CID 67298
IUPAC Name 2-chloroaniline;hydrochloride
SMILES [H+].[Cl-].NC1=CC=CC=C1Cl

Specifications

Melting Point ∼220°C (sublimation)
Odor Characteristic
Quantity 25 g
UN Number UN2018
Beilstein 3624280
Solubility Information It is soluble in water.
Formula Weight 164.03
Percent Purity 97%
Chemical Name or Material 2-Chloroaniline hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.