missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroaniline, 98+%

Product Code. 11412418
Change view
Click to view available options
Quantity:
10,000 g
2500 g
500 g
Unit Size:
10000g
2500g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11412418 10,000 g 10000g
11492408 500 g 500g
11402418 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11412418 Supplier Thermo Scientific Alfa Aesar Supplier No. A15758.0C
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 95-51-2 | C6H6ClN | 127.571 g/mol

2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroaniline used as petroleum solvents. It is used in biological studies to evaluate the mutagenicity and carcinogenicity of Salmonella typhimurium. Used as a Intermediates for dyestuff, pigment and specialty chemicals. 2-Chloroaniline is widely used in polymer, rubber, pharmaceutical and dye industries.

Solubility
Miscible with alcohol, ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).

Notes
Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.571
MDL Number MFCD00007656
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
Synonym o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
PubChem CID 7240
IUPAC Name 2-chloroaniline
SMILES C1=CC=C(C(=C1)N)Cl

Specifications

Melting Point -3°C
Density 1.213
Quantity 10,000 g
Odor Characteristic
Refractive Index 1.588
UN Number UN2019
Beilstein 606077
Merck Index 14,2118
Boiling Point 207°C to 209°C
Solubility Information Miscible with alcohol,ether. Soluble in acid and most organic solvents. Very slightly soluble in water (0.5g/100mL at 20°C).
Formula Weight 127.57
Percent Purity ≥98%
Chemical Name or Material 2-Chloroaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.