missing translation for 'onlineSavingsMsg'
Learn More

2-chloroaniline, 98+%

Product Code. 10050910 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mL
1 L
5 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
250mL
5 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10050910 1 L Glass bottle 1 L
10657252 250 mL Glass bottle 250mL
10676354 5 L Plastic drum 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10050910 Supplier Thermo Scientific Acros Supplier No. 108570010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 95-51-2 | C6H6ClN | 127.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 95-51-2
Molecular Formula C6H6ClN
Molecular Weight (g/mol) 127.57
MDL Number MFCD00007656
InChI Key AKCRQHGQIJBRMN-UHFFFAOYSA-N
Synonym o-chloroaniline, benzenamine, 2-chloro, 2-chlorobenzenamine, 2-chlorophenylamine, 1-amino-2-chlorobenzene, o-aminochlorobenzene, o-chloroaminobenzene, aniline, o-chloro, o-chloraniline, fast yellow gc base
PubChem CID 7240
IUPAC Name 2-chloroaniline
SMILES C1=CC=C(C(=C1)N)Cl

Specifications

Melting Point -3°C
CAS Min % 98.0
Color Yellow to Brown
CAS Max % 100.0
Density 1.2100g/mL
Quantity 1 L
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5870 to 1.589
Assay Percent Range 98% min. (GC)
Specific Gravity 1.21
Linear Formula ClC6H4NH2
Beilstein 12, 597
Merck Index 15, 2118
Boiling Point 209°C
Solubility Information Solubility in water: 5.13g/L (20°C). Other solubilities: soluble in most common organic solvents,miscible with alcohol and ether
Formula Weight 127.57
Percent Purity 98+%
Chemical Name or Material 2-Chloroaniline
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloroaniline
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H373-May cause damage to organs.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H301-Toxic if swallowed.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.