missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ 2-Chloroadenosine
Description
Metabolically stable analog of adenosine that behaves as an adenosine receptor agonist (Ki values are 300, 80 and 1900 nM for A1, A2A and A3 receptors respectively). Anticonvulsive in vivo.
Specifications
Specifications
| CAS | 146-77-0 |
| Quantity | 50 mg |
| Molecular Formula | C10H12ClN5O4 |
| Formula Weight | 301.69 |
| Purity | >98% |
| Molecular Weight (g/mol) | 301.69 |
| Chemical Name or Material | 2-Chloroadenosine |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?