missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-Chloroadenosine, Mikromol™
Description
2-Chloroadenosine
Specifications
Specifications
| Chemical Name or Material | 2-Chloro Adenosine |
| CAS | 146-77-0 |
| Analyte or Component Names | 2-Chloro Adenosine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C10 H12 Cl N5 O4 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Adenosine, 2-chloro-, 2-Chloroadenosine, 2-Chloro-D-adenosine, Antibiotic AT 265B, CADO, NSC 36896 |
| InChI Formula | InChI=1S/C10H12ClN5O4/c11-10-14-7(12)4-8(15-10)16(2-13-4)9-6(19)5(18)3(1-17)20-9/h2-3,5-6,9,17-19H,1H2,(H2,12,14,15)/t3-,5-,6-,9-/m1/s1 |
| SMILES | Nc1nc(Cl)nc2c1ncn2[C@@H]3O[C@H](CO)[C@@H](O)[C@H]3O |
| IUPAC Name | (2R,3R,4S,5R)-2-(6-amino-2-chloropurin-9-yl)-5-(hydroxymethyl)oxolane-3,4-diol |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?