missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacrylonitrile, 99%, stabilized

Product Code. 10157843 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10157843 25 g Glass bottle 25g
10623332 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10157843 Supplier Thermo Scientific Acros Supplier No. 108560250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 920-37-6 | C3H2ClN | 87.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 920-37-6
Molecular Formula C3H2ClN
Molecular Weight (g/mol) 87.51
MDL Number MFCD00001858
InChI Key OYUNTGBISCIYPW-UHFFFAOYSA-N
Synonym 2-chloroacrylonitrile, 2-propenenitrile, 2-chloro, chloroacrylonitrile, acrylonitrile, 2-chloro, 2-chloro-2-propenenitrile, unii-ffj1qhd154, ccris 6869, alpha-chloroacrylonitrile, .alpha.-chloroacrylonitrile, ffj1qhd154
PubChem CID 70198
IUPAC Name 2-chloroprop-2-enenitrile
SMILES ClC(=C)C#N

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -65°C
Color Colorless to Yellow
Density 1.0960g/mL
Boiling Point 88°C to 89°C
Flash Point 6°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula H2C=C(Cl)CN
Refractive Index 1.4290 to 1.431
Quantity 25 g
Fieser 03,66; 04,76; 05,107; 06,142; 08,444; 09,75; 11,139; 16,391
Specific Gravity 1.096
Stabilizer 500ppm (ca.) hydroquinone
Formula Weight 87.51
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-Chloroacrylonitrile, Stabilized
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 2-Chloroacrylonitrile, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H300-Fatal if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H351-Suspected of causing cancer.
  • H311-Toxic in contact with skin.
  • H331-Toxic if inhaled.
  • H335-May cause respiratory irritation.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.