missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetophenone, TRC

Product Code. 30350019
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30350019 25 g 25g
30349098 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30350019 Supplier Toronto Research Chemicals Supplier No. TRCC36381525G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-68-9
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.59
Synonym 1-(2-Chlorophenyl)ethanone, 2-Acetylphenyl Chloride, 2-Chloro-1-acetylbenzene, 2-Chlorophenyl Methyl Ketone, NSC 405474, o-Chloroacetophenone
IUPAC Name 1-(2-chlorophenyl)ethanone
SMILES CC(=O)c1ccccc1Cl

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H7ClO/c1-6(10)7-4-2-3-5-8(7)9/h2-5H,1H3
Formula Weight 154.0185
Chemical Name or Material 2'-Chloroacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.