missing translation for 'onlineSavingsMsg'
Learn More

2'-Chloroacetophenone, 98%

Product Code. 11452268
Change view
Click to view available options
Quantity:
50 g
250 g
1000 g
Unit Size:
1000g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11452268 250 g 250g
11442268 50 g 50g
11432268 1000 g 1000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11452268 Supplier Thermo Scientific Alfa Aesar Supplier No. A15232.30

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2142-68-9 | C8H7ClO | 154.593 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2142-68-9
Molecular Formula C8H7ClO
Molecular Weight (g/mol) 154.593
MDL Number MFCD00000560
InChI Key ZDOYHCIRUPHUHN-UHFFFAOYSA-N
Synonym 2'-chloroacetophenone, 1-2-chlorophenyl ethanone, o-chloroacetophenone, 1-2-chlorophenyl ethan-1-one, ethanone, 1-2-chlorophenyl, acetophenone, 2'-chloro, o-chloroacetophonone, 1-2-chloro-phenyl-ethanone, ethanone, 1-chlorophenyl, 1-acetyl-2-chlorobenzene
PubChem CID 72864
IUPAC Name 1-(2-chlorophenyl)ethanone
SMILES CC(=O)C1=CC=CC=C1Cl

Specifications

Density 1.189
Boiling Point 228°C to 229°C
Flash Point 88°C (190°F)
Refractive Index 1.5435
Quantity 250 g
UN Number UN3416
Beilstein 1858916
Formula Weight 154.6
Percent Purity 98%
Chemical Name or Material 2'-Chloroacetophenone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.