missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2'-Chloroacetanilide, Mikromol™
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
Description
2'-Chloroacetanilide, Mikromol
Specifications
Specifications
| Chemical Name or Material | 2'-Chloroacetanilide |
| CAS | 533-17-5 |
| Analyte or Component Names | 2'-Chloroacetanilide |
| Shipping Condition | Room Temperature |
| Molecular Formula | C8 H8 Cl N O |
| Certifications/Compliance | ISO 9001 |
| Synonym | Acetanilide, 2'-chloro- (8CI), Acetanilide, o-chloro- (3CI), 2'-Chloroacetanilide, N-(2-Chlorophenyl)acetamide, NSC 40562, NSC 8455, o-Chloroacetanilide |
| InChI Formula | InChI=1S/C8H8ClNO/c1-6(11)10-8-5-3-2-4-7(8)9/h2-5H,1H3,(H,10,11) |
| Solution Type | Neat |
| SMILES | CC(=O)Nc1ccccc1Cl |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction