missing translation for 'onlineSavingsMsg'
Learn More

2-Chloroacetamide, 98+%

Product Code. 11462278
Change view
Click to view available options
Quantity:
250 g
1000 g
5000 g
Unit Size:
1000g
250g
5000g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11462278 250 g 250g
11452278 1000 g 1000g
11472278 5000 g 5000g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11462278 Supplier Thermo Scientific Alfa Aesar Supplier No. A15238.30
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 79-07-2 | C2H4ClNO | 93.51 g/mol

2-Chloroacetamide is used as surface-active agent in home laundry, cleaning products & household formulated cleaners, other industrial uses, petroleum production, polymer processing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloroacetamide is used as surface-active agent in home laundry, cleaning products & household formulated cleaners, other industrial uses, petroleum production, polymer processing.

Solubility
Soluble in water(90g/L).

Notes
Store in cool, dry conditions in well sealed containers. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-07-2
Molecular Formula C2H4ClNO
Molecular Weight (g/mol) 93.51
MDL Number MFCD00008027
InChI Key VXIVSQZSERGHQP-UHFFFAOYSA-N
Synonym chloroacetamide, acetamide, 2-chloro, chloracetamide, 2-chloroethanamide, microcide, mergal af, chloracetamid, usaf do-29, 2-chloracetamide, 2-chloro-acetamide
PubChem CID 6580
IUPAC Name 2-chloroacetamide
SMILES NC(=O)CCl

Specifications

Melting Point 116°C to 119°C
Boiling Point 220°C (decomposition)
Flash Point 170°C (338°F)
Odor Weak
Quantity 250 g
UN Number UN2811
Beilstein 878191
Merck Index 14,2110
Solubility Information Soluble in water(90g/L).
Formula Weight 93.51
Percent Purity ≥98%
Chemical Name or Material 2-Chloroacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloroacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Reproductive toxicity Category 2
  • Skin sensitiser Category 1
Hazard Statement
  • H301-Toxic if swallowed.
  • H317-May cause an allergic skin reaction.
  • H361f-Suspected of damaging fertility.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P405-Store locked up.
Supplemental information
  • MIXTURE LIST-Contain: 2-Chloroacetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.