missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-phenylacetamide, TRC

Product Code. 30357668
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357668 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357668 Supplier Toronto Research Chemicals Supplier No. TRCC3753701G
 Restricted Item
Estimated Shipment: 06-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-N-phenylacetamide
CAS 587-65-5
Molecular Formula C8H8ClNO
Quantity 1 g
Synonym 2-Chloroacetanilide, Chloracetanilide, Chloroacetanilide, N-(2-Chloroacetyl)aniline, N-(Chloroacetyl)aniline, N-Phenyl-2-chloroacetamide, N-Phenyl-2-chloroethanamide, N-Phenylchloroacetamide, NSC 311, Phenylcarbamoylmethyl Chloride, alpha-Chloro-N-phenylacetamide, alpha-Chloroacetanilide, ω-Chloroacetanilide
InChI Formula InChI=1S/C8H8ClNO/c9-6-8(11)10-7-4-2-1-3-5-7/h1-5H,6H2,(H,10,11)
SMILES ClCC(=O)Nc1ccccc1
IUPAC Name 2-chloro-N-phenylacetamide
Molecular Weight (g/mol) 169.03
Formula Weight 169.0294
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.