missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-dimethylbenzamide, 97%, Thermo Scientific Chemicals

Product Code. 15446408
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15446408 1 g 1g
15456408 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446408 Supplier Thermo Scientific Alfa Aesar Supplier No. H59948.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Chloro-N,N-dimethylbenzamide is involved in the preparation of repellents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Chloro-N,N-dimethylbenzamide is involved in the preparation of repellents.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6526-67-6
Molecular Formula C9H10ClNO
Molecular Weight (g/mol) 183.64
MDL Number MFCD00463636
InChI Key SZUQPOLAEFBJJH-UHFFFAOYSA-N
Synonym benzamide, 2-chloro-n,n-dimethyl, n,n-dimethyl-2-chlorobenzamide, o-chloro-n,n-dimethylbenzamide, 2-chloro-n,n-dimethyl-benzamide, benzamide, o-chloro-n,n-dimethyl, aronis25704, 2-chlorophenyl-n,n-dimethylcarboxamide
PubChem CID 81012
IUPAC Name 2-chloro-N,N-dimethylbenzamide
SMILES CN(C)C(=O)C1=CC=CC=C1Cl

Specifications

Color Yellow
Quantity 1 g
Formula Weight 183.64
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-Chloro-N,N-dimethylbenzamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.