missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N,N-dimethylacetamide, 97%

Product Code. 11479340
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11479340 250 mg 250mg
11489340 1 g 1g
15417867 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11479340 Supplier Thermo Scientific Alfa Aesar Supplier No. H50229.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2675-89-0 | C4H8ClNO | 121.564 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2675-89-0
Molecular Formula C4H8ClNO
MDL Number MFCD00039360
InChI Key XBPPLECAZBTMMK-UHFFFAOYSA-N
PubChem CID 75886
IUPAC Name 2-chloro-N,N-dimethylacetamide
SMILES CN(C)C(=O)CCl

Specifications

Density 1.182
Boiling Point 98°C to 100°C (11 mmHg)
Linear Formula ClCH2CON(CH3)2
Refractive Index 1.479
Quantity 250 mg
Beilstein 969568
Synonym n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide, acetamide, 2-chloro-n,n-dimethyl, 2-chloro-n, n-dimethylacetamide, acmc-209gsx, 2-chlorodimethylacetamide, 2-chloro-dimethylacetamide, 2-chlorodimethyl acetamide, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide
Molecular Weight (g/mol) 121.564
Formula Weight 121.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-N,N-dimethylacetamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.