missing translation for 'onlineSavingsMsg'
Learn More

2-chloro-N,N-dimethylacetamide, 97%

Product Code. 10027899 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10027899 5 mL Glass bottle 5mL
10422010 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10027899 Supplier Thermo Scientific Acros Supplier No. 438210050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 2675-89-0 | C4H8ClNO | 121.57 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2675-89-0
Molecular Formula C4H8ClNO
Molecular Weight (g/mol) 121.57
InChI Key XBPPLECAZBTMMK-UHFFFAOYSA-N
Synonym n,n-dimethyl-2-chloroacetamide, n,n-dimethylchloroacetamide, acetamide, 2-chloro-n,n-dimethyl, 2-chloro-n, n-dimethylacetamide, acmc-209gsx, 2-chlorodimethylacetamide, 2-chloro-dimethylacetamide, 2-chlorodimethyl acetamide, chloro-n,n-dimethylacetamide, n,n-dimethyl chloroacetamide
PubChem CID 75886
IUPAC Name 2-chloro-N,N-dimethylacetamide
SMILES CN(C)C(=O)CCl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 15°C to 16°C
Color Colorless to Yellow
Density 1.1820g/mL
Boiling Point 98°C to 100°C (11.0 Torr)
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula ClCH2CON(CH3)2
Quantity 5 mL
Specific Gravity 1.182
Solubility Information Solubility in water: in water:(20°C) soluble.,in ethanol:(20°C) soluble
Formula Weight 121.57
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2-chloro-N, N-dimethylacetamide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 2-Chloro-N,N-dimethylacetamide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.