missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methylacetamide, 97%

Product Code. 15405087
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 15405087

Brand: Thermo Scientific Alfa Aesar H32362.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 96-30-0 | C3H6ClNO | 107.537 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96-30-0
Molecular Formula C3H6ClNO
Molecular Weight (g/mol) 107.537
MDL Number MFCD00018913
InChI Key HOZLOOPIXHWKCI-UHFFFAOYSA-N
Synonym n-methyl-2-chloroacetamide, acetamide, 2-chloro-n-methyl, n-methylchloroacetamide, usaf do-35, 2-chloro-n-methyl-acetamide, alpha-chloro-n-methylacetamide, n-methyl 2-chloroacetamide, .alpha.-chloro-n-methylacetamide, n-methylchloracetamide, n-methyl chloroacetamide
PubChem CID 66773
IUPAC Name 2-chloro-N-methylacetamide
SMILES CNC(=O)CCl

Specifications

Melting Point 45°C to 47°C
Linear Formula ClCH2CONHCH3
Quantity 5 g
Formula Weight 107.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-methylacetamide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.