Learn More
2-Chloro-N-methoxy-N-methylacetamide, 98%
Description
2-Chloro-N-methoxy-N-methylacetamide is used in the preparation of 2-heptyl-3-hydroxy-4(1H)-quinolone (Pseudomonas quinolone signal or PQS) and structurally related 2-alkyl-4-quinolones having biological activity, 2-(benzo[d]thiazol-2-ylsulfonyl)-N-methoxy-N-methylacetamide and α-chloro-ketone, starting reagent for the one-pot synthesis of 2-heptyl-3-hydroxyl-4(1H)-quinolone (PQS), signaling molecule in the quorum sensing of Pseudomonas aeruginosa.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
2-Chloro-N-methoxy-N-methylacetamide is used in the preparation of 2-heptyl-3-hydroxy-4(1H)-quinolone (Pseudomonas quinolone signal or PQS) and structurally related 2-alkyl-4-quinolones having biological activity, 2-(benzo[d]thiazol-2-ylsulfonyl)-N-methoxy-N-methylacetamide and α-chloro-ketone, starting reagent for the one-pot synthesis of 2-heptyl-3-hydroxyl-4(1H)-quinolone (PQS), signaling molecule in the quorum sensing of Pseudomonas aeruginosa.
Solubility
Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2-8°C. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 39°C to 41°C |
| Boiling Point | 94°C to 95°C (9 mmHg) |
| Flash Point | 104°C (219°F) |
| Quantity | 25 g |
| Beilstein | 1924015 |
| Synonym | 2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide |
| Solubility Information | Insoluble in water. |
| Formula Weight | 137.57 |
| Percent Purity | 98% |
| Chemical Name or Material | 2-Chloro-N-methoxy-N-methylacetamide |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.