missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-methoxy-N-methylacetamide, 97%, Thermo Scientific™

Product Code. 10212672 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10212672 5 g Glass bottle 5g
10458172 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10212672 Supplier Thermo Scientific Acros Supplier No. 369890050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67442-07-3
MDL Number MFCD00134232
InChI Key SCOJKGRNQDKFRP-UHFFFAOYSA-N
Synonym 2-chloro-n-methoxy-n-methyl acetamide, acetamide,2-chloro-n-methoxy-n-methyl, n-methoxy-n-methylchloroacetamide, acetamide, 2-chloro-n-methoxy-n-methyl, pubchem18657, acmc-209nz0, n-methyl-n-methoxy chloroacetamide, n-chloroacetyl-n-methoxymethylamine, n-methyl-n-methoxy-2-chloroacetamide, 2-chloro-n-methoxy-n-methyl-acetamide
PubChem CID 2734716
IUPAC Name 2-chloro-N-methoxy-N-methylacetamide
SMILES CN(C(=O)CCl)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 37.0°C to 42.0°C
Color White to Yellow
Boiling Point 94.0°C to 95.0°C
Flash Point 104°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C4H8ClNO2
Linear Formula ClCH2CON(OCH3)CH3
Quantity 5 g
Molecular Weight (g/mol) 137.56
Formula Weight 137.56
Percent Purity 97%
Physical Form Crystals
Chemical Name or Material 2-Chloro-N-methoxy-N-methylacetamide
Show More Show Less
compliance-icons
Product Identifier
  • 2-Chloro-N-methoxy-N-methylacetamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.