missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-(hydroxymethyl)acetamide, 98%

Product Code. 11494943
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11494943 50 g 50g
11484943 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11494943 Supplier Thermo Scientific Alfa Aesar Supplier No. A12247.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2832-19-1 | C3H6ClNO2 | 123.54 g/mol

An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
An organic building block. It is an important raw material and intermediate used in Organic Synthesis, Pharmaceuticals, Agrochemicals. As dyestuff intermediate .

Solubility
Very soluble in water. Poorly soluble in aliphatic hydrocarbons.

Notes
Hygroscopic. Store in cool, dry conditions in well sealed containers. Store away from strong oxidizing agents. Keep container tightly closed.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2832-19-1
Molecular Formula C3H6ClNO2
Molecular Weight (g/mol) 123.54
MDL Number MFCD00021961
InChI Key TXNSZCSYBXHETP-UHFFFAOYSA-N
Synonym 2-chloro-n-hydroxymethyl acetamide, n-hydroxymethyl-2-chloroacetamide, acetamide, 2-chloro-n-hydroxymethyl, n-methylolchloroacetamide, grota, chloracetamide-n-metholol, n-methylol-chloracetamide, grotan df-35, caswell no. 192bb, chloracetamide-n-methanol
PubChem CID 62466
IUPAC Name 2-chloro-N-(hydroxymethyl)acetamide
SMILES OCNC(=O)CCl

Specifications

Melting Point 97°C to 103°C
Quantity 50 g
UN Number UN3261
Beilstein 1811765
Sensitivity Hygroscopic
Solubility Information Very soluble in water. Poorly soluble in aliphatic hydrocarbons.
Formula Weight 123.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-N- (hydroxymethyl)acetamide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.