missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-N-ethyl-N-(3-methylphenyl)benzamide, 97%, Thermo Scientific™

Catalog Number 15485196
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
1018134-12-7
C16H16ClNO
273.76
MFCD22683085
YFBNVKAYRHEGFA-UHFFFAOYSA-N
2-chloro-n-ethyl-n-3-methylphenyl benzamide
39847595
2-chloro-N-ethyl-N-(3-methylphenyl)benzamide
CCN(C(=O)C1=CC=CC=C1Cl)C1=CC=CC(C)=C1
This item is not returnable. View return policy
1018134-12-7
C16H16ClNO
273.76
MFCD22683085
YFBNVKAYRHEGFA-UHFFFAOYSA-N
2-chloro-n-ethyl-n-3-methylphenyl benzamide
39847595
2-chloro-N-ethyl-N-(3-methylphenyl)benzamide
CCN(C(=O)C1=CC=CC=C1Cl)C1=CC=CC(C)=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Formula Weight 273.76
Percent Purity 97%
Chemical Name or Material 2-Chloro-N-ethyl-N-(3-methylphenyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'