missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-m-xylene, 98%

Product Code. 11473468
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11473468 25 g 25g
11483468 100 g 100g
11493468 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11473468 Supplier Thermo Scientific Alfa Aesar Supplier No. A17867.14
 Restricted Item
Estimated Shipment: 04-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 6781-98-2 | C8H9Cl | 140.61 g/mol

It is employed as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed as intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in a tightly closed container in a cool, dry, well-ventilated area. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6781-98-2
Molecular Formula C8H9Cl
Molecular Weight (g/mol) 140.61
MDL Number MFCD00000565
InChI Key VDXLAYAQGYCQEO-UHFFFAOYSA-N
Synonym 2-chloro-m-xylene, 2,6-dimethylchlorobenzene, m-xylene, 2-chloro, benzene, 2-chloro-1,3-dimethyl, 2,6-dimethyl chlorobenzene, 2,6-dimethylphenyl chloride, 1-chloro-2,6-dimethylbenzene, 2,6-dimethylchlorobenzene;2-chloro-m-xylene, 2-chloro-meta-xylene, pubchem3643
PubChem CID 32885
IUPAC Name 2-chloro-1,3-dimethylbenzene
SMILES CC1=C(C(=CC=C1)C)Cl

Specifications

Melting Point -36°C
Density 1.061
Quantity 25 g
Refractive Index 1.527
Beilstein 2040954
Boiling Point 182°C to 183°C
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 140.61
Percent Purity 98%
Chemical Name or Material 2-Chloro-m-xylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.