missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-L-phenylalanine, 98+%

Product Code. p-7029646
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15463617 250 mg 250mg
11486240 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15463617 Supplier Thermo Scientific Alfa Aesar Supplier No. H27135.MD
Add to basket
This item is not returnable. View return policy

CAS: 103616-89-3 | C9H10ClNO2 | 199.63 g/mol

The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
The largest gain in affinity of all tested unnatural amino acids was observed for 2-chloro-L-phenylalanine.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103616-89-3
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD00077921
InChI Key CVZZNRXMDCOHBG-QMMMGPOBSA-N
Synonym 2-chloro-l-phenylalanine, l-2-chlorophenylalanine, s-2-amino-3-2-chlorophenyl propanoic acid, h-phe 2-cl-oh, 2s-2-amino-3-2-chlorophenyl propanoic acid, phenylalanine, 2-chloro, l-2-chlorophe, l-2-chloro phenylalanine, s-2-chlorophenylalanine, ksc491q0l
PubChem CID 2761491
IUPAC Name (2S)-2-amino-3-(2-chlorophenyl)propanoic acid
SMILES N[C@@H](CC1=C(Cl)C=CC=C1)C(O)=O

Specifications

Melting Point 196°C to 199°C
Solubility Information Insoluble in water.
Quantity 250 mg
Formula Weight 199.64
Percent Purity ≥98%
Chemical Name or Material 2-Chloro-L-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.