missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro Fenofibric Acid, TRC

Product Code. 30353044
Change view
Click to view available options
Quantity:
25 mg
100 mg
250 mg
Unit Size:
250mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30353044 250 mg 250mg
30364477 25 mg 25mg
30364164 100 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30353044 Supplier Toronto Research Chemicals Supplier No. TRCC364675250MG
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61024-31-5
Molecular Formula C17H15ClO4
Molecular Weight (g/mol) 318.75
Synonym 2-[4-(2-Chlorobenzoyl)phenoxy]-2-methylpropanoic Acid, LF 590
IUPAC Name 2-[4-(2-chlorobenzoyl)phenoxy]-2-methylpropanoic acid
SMILES CC(C)(Oc1ccc(cc1)C(=O)c2ccccc2Cl)C(=O)O

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C17H15ClO4/c1-17(2,16(20)21)22-12-9-7-11(8-10-12)15(19)13-5-3-4-6-14(13)18/h3-10H,1-2H3,(H,20,21)
Formula Weight 318.0659
Chemical Name or Material 2-Chloro Fenofibric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.