missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-9-methyl-6-(benzylamino)purine (>90%), TRC

Product Code. 30352934
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30352934 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30352934 Supplier Toronto Research Chemicals Supplier No. TRCC368860250MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-Chloro-9-methyl-6-(benzylamino)purine
CAS 101622-53-1
Molecular Formula C13H12ClN5
Quantity 250 mg
Synonym 6-(Benzylamino)-2-chloro-9-methylpurine, 2-Chloro-6-benzylamino-9-methylpurine, 2-Chloro-9-methyl-N-(phenylmethyl)-9H-purin-6-amine
InChI Formula InChI=1S/C13H12ClN5/c1-19-8-16-10-11(17-13(14)18-12(10)19)15-7-9-5-3-2-4-6-9/h2-6,8H,7H2,1H3,(H,15,17,18)
SMILES CCc1ccc(cc1)OCC2OC2
IUPAC Name N-benzyl-2-chloro-9-methylpurin-6-amine
Molecular Weight (g/mol) 273.72
Product Title
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.