missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-6-(trifluoromethyl)benzamide, 97%, Thermo Scientific™

Product Code. 15405247
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15405247 1 g 1g
15415247 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15405247

Brand: Thermo Scientific Alfa Aesar H32515.03

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 916420-45-6
Molecular Formula C8H5ClF3NO
Molecular Weight (g/mol) 223.579
MDL Number MFCD09025362
InChI Key JUAICIUGQMFFPO-UHFFFAOYSA-N
Synonym 2-chloro-6-trifluoromethyl benzamide, 6-chloro-2-trifluoromethyl benzamide
PubChem CID 46737494
IUPAC Name 2-chloro-6-(trifluoromethyl)benzamide
SMILES C1=CC(=C(C(=C1)Cl)C(=O)N)C(F)(F)F

Specifications

Melting Point 155°C to 158°C
Quantity 1 g
Formula Weight 223.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-6-(trifluoromethyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.